Assay Details
Target Gene Details
Entrez Gene ID: | 20592 |
Gene Name: | lysine (K)-specific demethylase 5D |
Gene Aliases: |
HY, Jarid1d, Smcy |
Location: |
Chr.Y:897660-943813 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm5d | NM_011419.3 | 14 | 2134 | NP_035549.1 |
| AB221607.1 | 14 | 1960 | ||
| AF127244.1 | 14 | 1972 | AAD53048.1 | |
| AK140971.1 | 14 | 2129 | BAE24534.1 | |
| BC059077.1 | 6 | 949 | AAH59077.1 |
More Information
Set Membership: |
|

Genomic Map