Assay Details
Target Gene Details
Entrez Gene ID: | 11798 |
Gene Name: | X-linked inhibitor of apoptosis |
Gene Aliases: |
1110015C02Rik, Aipa, Api3, Birc4, IAP3, ILP-1, MIHA |
Location: |
Chr.X:42060402-42109664 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xiap | NM_001301639.1 | NP_001288568.1 | ||
| NM_001301641.1 | NP_001288570.1 | |||
| NM_009688.3 | NP_033818.2 | |||
| NR_125870.1 | ||||
| AK147447.1 | ||||
| BB650856.1 | ||||
| BB663325.1 | ||||
| BC138528.1 | ||||
| BC145861.1 | ||||
| U36842.1 | AAC52594.1 | |||
| U88990.1 | AAB58376.1 |
More Information
Set Membership: |
|

Genomic Map