Assay Details
Target Gene Details
Entrez Gene ID: | 320718 |
Gene Name: | solute carrier family 26, member 9 |
Gene Aliases: |
E030002L01Rik |
Location: |
Chr.1:131744022-131771504 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a9 | NM_177243.4 | 21 | 3036 | NP_796217.2 |
| AK078966.1 | BAC37482.1 | |||
| AK086815.1 | 21 | 3037 | BAC39749.1 | |
| AY034145.1 | 20 | 2949 | AAK54448.1 |
More Information
Set Membership: |
|

Genomic Map