Assay Details
Target Gene Details
Entrez Gene ID: | 12043 |
Gene Name: | B cell leukemia/lymphoma 2 |
Gene Aliases: |
AW986256, Bcl-2, C430015F12Rik, D630044D05Rik, D830018M01Rik |
Location: |
Chr.1:106538178-106714290 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2 | NM_009741.4 | 2 | 4592 | NP_033871.2 |
| AK052879.1 | 1 | 1144 | ||
| AK076269.1 | 1 | 117 | ||
| AK155394.1 | 1 | 894 | ||
| BB612176.1 | 1 | 115 |

Genomic Map