Assay Details
Target Gene Details
Entrez Gene ID: | 246229 |
Gene Name: | basic, immunoglobulin-like variable motif containing |
Gene Aliases: |
AI854267 |
Location: |
Chr.1:44118888-44144771 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bivm | NM_144558.4 | 1 | 251 | NP_653141.3 |
| AF411386.1 | AAM45762.1 | |||
| AK032375.1 | 1 | 213 | BAC27843.1 | |
| AK034329.1 | 1 | 213 | BAC28676.1 | |
| AK047404.1 | 1 | 253 | ||
| AK158410.1 | 1 | 247 | ||
| BC082584.1 | AAH82584.1 |
More Information
Set Membership: |
|

Genomic Map