Assay Details
Target Gene Details
Entrez Gene ID: | 226866 |
Gene Name: | somatomedin B and thrombospondin, type 1 domain containing |
Gene Aliases: |
Gm106, Rpesp |
Location: |
Chr.1:15853862-15892722 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sbspon | NM_001033288.3 | 5 | 2097 | NP_001028460.1 |
| AK137144.1 | 5 | 2098 | BAE23250.1 |

Genomic Map