Assay Details
Target Gene Details
Entrez Gene ID: | 16560 |
Gene Name: | kinesin family member 1A |
Gene Aliases: |
ATSV, C630002N23Rik, Gm1626, Kns1 |
Location: |
Chr.1:93015455-93101890 on Build GRCm38 |
Assay Gene Location: | Within Exon 50 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif1a | NM_001110315.2 | 47 | 6215 | NP_001103785.1 |
| NM_001294149.1 | 48 | 6242 | NP_001281078.1 | |
| NM_001294150.1 | 47 | 6130 | NP_001281079.1 | |
| NM_008440.4 | 48 | 6239 | NP_032466.2 | |
| AK083095.1 | 1 | 739 | ||
| AK220487.1 | 32 | 4391 | BAD90508.1 |
More Information
Set Membership: |
|

Genomic Map