Assay Details
Target Gene Details
Entrez Gene ID: | 227059 |
Gene Name: | solute carrier family 39 (zinc transporter), member 10 |
Gene Aliases: |
2900042E17Rik, 5430433I10, mKIAA1265 |
Location: |
Chr.1:46807544-46925772 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a10 | NM_172653.2 | 7 | 1820 | NP_766241.2 |
| AK049099.1 | 5 | 566 | BAC33542.1 | |
| AK122483.1 | 6 | 1254 | BAC65765.1 | |
| AK136117.1 | 6 | 1532 | ||
| BC052880.1 | 7 | 1934 | AAH52880.1 | |
| BC059214.1 | 5 | 704 | AAH59214.1 | |
| BC062918.1 | 7 | 1820 | AAH62918.1 |
More Information
Set Membership: |
|

Genomic Map