Assay Details
Target Gene Details
Entrez Gene ID: | 17974 |
Gene Name: | non-catalytic region of tyrosine kinase adaptor protein 2 |
Gene Aliases: |
4833426I10Rik, Grb4, NCKbeta, Nck-2 |
Location: |
Chr.1:43445749-43570518 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nck2 | NM_010879.3 | 1 | 182 | NP_035009.3 |
| AK014772.1 | ||||
| AK014949.1 | 1 | 176 | ||
| AK051665.1 | 1 | 182 | BAC34709.1 |
More Information
Set Membership: |
|

Genomic Map