Assay Details
Target Gene Details
Entrez Gene ID: | 72481 |
Gene Name: | RIKEN cDNA 2610203C22 gene |
Gene Aliases: |
2310007M24Rik |
Location: |
Chr.1:9560832-9631092 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610203C22Rik | NR_015470.1 | |||
| AK011866.1 |
Target Gene Details
Entrez Gene ID: | 76187 |
Gene Name: | alcohol dehydrogenase, iron containing, 1 |
Gene Aliases: |
6330565B14Rik, AI043035, Adh8 |
Location: |
Chr.1:9547969-9577968 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adhfe1 | NM_175236.4 | 14 | 1653 | NP_780445.1 |
| NR_027664.1 | 13 | 1612 | ||
| AK018241.1 | 14 | 1624 | ||
| AK038853.1 | 14 | 1620 | BAC30151.1 | |
| AK046780.1 | 14 | 1579 | ||
| AK050178.1 | 13 | 1577 | BAC34108.1 | |
| BC026584.1 | 14 | 1611 | AAH26584.2 |
More Information
Set Membership: |
|

Genomic Map