Assay Details
Target Gene Details
Entrez Gene ID: | 12902 |
Gene Name: | complement receptor 2 |
Gene Aliases: |
C3DR, CD21, CD35, Cr-1, Cr-2, Cr1 |
Location: |
Chr.1:195136811-195177190 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cr2 | NM_007758.2 | 13 | 2382 | NP_031784.1 |
| AK003072.2 | 13 | 2382 | BAB22548.1 | |
| BC109344.1 | 13 | 2315 | AAI09345.1 | |
| BC109345.1 | AAI09346.1 | |||
| M29281.1 | 8 | 1386 | AAA37447.1 | |
| M35684.1 | 12 | 2250 | AAA37448.1 | |
| M61132.1 | 12 | 2217 | AAA63295.1 |

Genomic Map