Assay Details
Target Gene Details
Entrez Gene ID: | 107508 |
Gene Name: | glutamyl-prolyl-tRNA synthetase |
Gene Aliases: |
2410081F06Rik, 3010002K18Rik, C79379, Qprs |
Location: |
Chr.1:185363028-185428355 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eprs | NM_029735.1 | 20 | 2925 | NP_084011.1 |
| AK028687.1 | 13 | 1946 | ||
| AK141072.1 | 20 | 2980 | ||
| AK148463.1 | 20 | 2886 | BAE28568.1 | |
| AK160986.1 | 20 | 2945 | ||
| BC040802.1 | 20 | 2923 | AAH40802.1 | |
| BC094679.1 | 19 | 2719 | AAH94679.1 | |
| BC141049.1 | 20 | 2844 | ||
| X54327.1 | 11 | 1609 |
More Information
Set Membership: |
|

Genomic Map