Assay Details
Target Gene Details
Entrez Gene ID: | 98685 |
Gene Name: | tRNA methyltransferase 1 like |
Gene Aliases: |
1190005F20Rik, AW226554, C1orf25, Trm1-like, Trm1l |
Location: |
Chr.1:151428547-151458183 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt1l | NM_026876.3 | 15 | 2249 | NP_081152.2 |
| AK004493.1 | 7 | 742 | BAB23331.1 | |
| AK146088.1 | 15 | 2250 | BAE26891.1 | |
| BC022682.1 | 10 | 1285 | AAH22682.1 | |
| BC037478.1 | 11 | 1448 | AAH37478.1 | |
| BC110425.1 | 15 | 2232 | ||
| BC132299.1 | 15 | 2107 |
More Information
Set Membership: |
|

Genomic Map