Assay Details
Target Gene Details
Entrez Gene ID: | 66875 |
Gene Name: | SWT1 RNA endoribonuclease homolog (S. cerevisiae) |
Gene Aliases: |
1200016B10Rik, C1orf26 |
Location: |
Chr.1:151367698-151428450 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Swt1 | NM_025819.4 | 6 | 1035 | NP_080095.3 |
| AK004803.2 | 6 | 982 | BAB23575.2 | |
| BC055064.1 | 6 | 1036 | AAH55064.1 | |
| BC060204.1 | 6 | 975 | AAH60204.1 |

Genomic Map