Assay Details
Target Gene Details
Entrez Gene ID: | 240832 |
Gene Name: | torsin A interacting protein 2 |
Gene Aliases: |
1110020D10Rik, 15kDa, A130072J07, AA103493, AW060462, AW610675, C77739, Ifrg15, Lull1 |
Location: |
Chr.1:156035664-156068861 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip2 | NM_001160180.1 | 3 | 1480 | NP_001153652.1 |
| NM_001160181.1 | 3 | 1428 | NP_001153653.1 | |
| NM_001160182.1 | 3 | 1359 | NP_001153654.1 | |
| NM_022329.4 | 2 | 1368 | NP_071724.1 | |
| NM_172843.4 | NP_766431.3 | |||
| AJ251363.1 | 2 | 1335 | CAC13975.1 | |
| AK003852.1 | 2 | 1317 | BAB23037.1 | |
| AK032697.1 | 2 | 1313 | BAC27991.1 | |
| AK038024.1 | BAC29923.1 | |||
| AK075980.1 | 1 | 599 | BAC36093.1 | |
| AK080959.1 | BAC38096.1 | |||
| AK150117.1 | 3 | 1483 | BAE29320.1 | |
| AK150303.1 | 3 | 1397 | BAE29454.1 | |
| AK151453.1 | 3 | 1397 | BAE30413.1 | |
| AK152137.1 | 3 | 1404 | BAE30976.1 | |
| BC064017.1 | 2 | 1333 | AAH64017.1 | |
| BC066790.1 | AAH66790.1 | |||
| BC092059.1 | 3 | 1386 | AAH92059.1 | |
| CF164425.1 |

Genomic Map