Assay Details
Target Gene Details
Entrez Gene ID: | 65102 |
Gene Name: | Ngg1 interacting factor 3-like 1 (S. pombe) |
Gene Aliases: |
1110030G24Rik |
Location: |
Chr.1:58445156-58462276 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nif3l1 | NM_022988.2 | 1 | 233 | NP_075364.2 |
| AF284439.1 | 2 | 710 | AAG45961.1 | |
| AK003978.1 | 2 | 340 | BAB23102.1 | |
| AK011670.1 | 2 | 250 | BAB27769.1 | |
| AK131631.1 | 2 | 596 | BAE20729.1 | |
| AK154815.1 | 2 | 325 | BAE32848.1 | |
| AK158616.1 | 2 | 616 | BAE34583.1 | |
| BC065163.1 | 1 | 233 | AAH65163.1 | |
| BC089475.1 | 2 | 253 | AAH89475.1 |

Genomic Map