Assay Details
Target Gene Details
Entrez Gene ID: | 110957 |
Gene Name: | DNA segment, Chr 1, Pasteur Institute 1 |
Gene Aliases: |
Pl10 |
Location: |
Chr.1:186967416-186970627 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D1Pas1 | NM_033077.3 | 1 | 539 | NP_149068.1 |
| AK029542.1 | 1 | 539 | BAC26505.1 | |
| BC139286.1 | 1 | 534 | ||
| BC139287.1 | 1 | 534 | ||
| J04847.1 | 1 | 79 | AAA39942.1 |

Genomic Map