Assay Details
Target Gene Details
Entrez Gene ID: | 67698 |
Gene Name: | family with sequence similarity 174, member A |
Gene Aliases: |
2310044D20Rik, Fam174, Naa6, Tmem157 |
Location: |
Chr.1:95313628-95335284 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam174a | NM_026321.4 | 1 | 517 | NP_080597.2 |
| AB154249.1 | 1 | 328 | ||
| AK009797.1 | 1 | 492 | BAB26509.1 | |
| AK009892.1 | 1 | 456 | BAB26567.1 | |
| AK017318.1 | 1 | 490 | BAB30690.1 | |
| AK020633.1 | 1 | 453 | BAB32159.1 | |
| AK033948.1 | 1 | 402 | BAC28523.1 | |
| AK084861.1 | 1 | 517 | ||
| AK148807.1 | 1 | 418 | BAE28669.1 | |
| BC021428.1 | 1 | 532 | AAH21428.1 |

Genomic Map