Assay Details
Target Gene Details
Entrez Gene ID: | 619288 |
Gene Name: | family with sequence similarity 71, member A |
Gene Aliases: |
4933417M04Rik, GARI-L4 |
Location: |
Chr.1:191162584-191164817 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam71a | NM_001109759.1 | 1 | 1112 | NP_001103229.1 |
| AB473728.1 | 1 | 739 | ||
| AK133412.1 | 1 | 1113 |

Genomic Map