Assay Details
Target Gene Details
Entrez Gene ID: | 72481 |
Gene Name: | RIKEN cDNA 2610203C22 gene |
Gene Aliases: |
2310007M24Rik |
Location: |
Chr.1:9560832-9631092 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610203C22Rik | NR_015470.1 | |||
| AK009214.1 | ||||
| AK011866.1 | ||||
| AK082741.1 |
Target Gene Details
Entrez Gene ID: | 76982 |
Gene Name: | RIKEN cDNA 3110035E14 gene |
Gene Aliases: |
AI415019, AV164974, R75066 |
Location: |
Chr.1:9601267-9627142 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110035E14Rik | NM_178399.4 | 6 | 2784 | NP_848486.1 |
| AK014123.1 | 1 | 1301 | ||
| AK043576.1 | 6 | 2631 | BAC31586.1 | |
| AK043759.1 | 6 | 2785 | BAC31645.1 | |
| BC066997.1 | 6 | 2546 | AAH66997.1 |
More Information
Set Membership: |
|

Genomic Map