Assay Details
Target Gene Details
Entrez Gene ID: | 52231 |
Gene Name: | ankyrin repeat and zinc finger domain containing 1 |
Gene Aliases: |
1300008P06Rik, 2810025E10Rik, D1Ertd161e |
Location: |
Chr.1:75192160-75199387 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankzf1 | NM_026187.5 | 9 | 1961 | NP_080463.3 |
| NR_051998.1 | 9 | 1992 | ||
| AK004948.2 | 7 | 2391 | ||
| AK012672.1 | 2 | 534 | BAB28398.1 | |
| AK012809.1 | 1 | 1105 | ||
| AK156255.1 | 9 | 1923 | BAE33643.1 | |
| AK157405.1 | 9 | 1923 | ||
| AK183698.1 | ||||
| AK207603.1 | ||||
| AW536302.1 | ||||
| BC057896.1 | 4 | 803 | AAH57896.1 | |
| BC150879.1 | 9 | 1911 |

Genomic Map