Assay Details
Target Gene Details
Entrez Gene ID: | 16980 |
Gene Name: | leucine rich repeat protein 2, neuronal |
Gene Aliases: |
5730406J09Rik, NLRR-2 |
Location: |
Chr.1:132880355-132940005 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrn2 | NM_010732.4 | 2 | 1310 | NP_034862.1 |
| AK036271.1 | 2 | 1311 | ||
| AK039216.1 | 2 | 1311 | ||
| AY362823.1 | 1 | 899 | AAQ74241.1 | |
| BC056458.1 | 2 | 1052 | AAH56458.1 | |
| D49375.1 | BAA08373.1 |
More Information
Set Membership: |
|

Genomic Map