Assay Details
Target Gene Details
Entrez Gene ID: | 71111 |
Gene Name: | G protein-coupled receptor 39 |
Gene Aliases: |
4933415E13Rik |
Location: |
Chr.1:125676996-125873862 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr39 | NM_027677.2 | 2 | 1381 | NP_081953.2 |
| AK016817.1 | 2 | 509 | BAB30446.1 | |
| BC085285.1 | 2 | 1394 | AAH85285.1 |
Target Gene Details
Entrez Gene ID: | 72585 |
Gene Name: | Ly6/Plaur domain containing 1 |
Gene Aliases: |
2700050C12Rik, AI853408, C530008O16Rik, Lynx2, Lypdc1 |
Location: |
Chr.1:125867635-125913186 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lypd1 | NM_145100.3 | 3 | 1533 | NP_659568.2 |
| AB041649.1 | 3 | 1533 | BAA95101.1 | |
| AK012406.1 | 3 | 1221 | BAE43232.1 | |
| AK045592.1 | 3 | 1534 | BAC32428.1 | |
| AK082941.1 | 3 | 1213 | BAE43380.1 | |
| BC058599.1 | 3 | 1466 | AAH58599.1 | |
| BX630278.1 |
More Information
Set Membership: |
|

Genomic Map