Assay Details
Target Gene Details
Entrez Gene ID: | 226970 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 4 |
Gene Aliases: |
9330140K16Rik, Asef, ENSMUSG00000070955 |
Location: |
Chr.1:34801722-34812754 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef4 | NM_183019.2 | 10 | 1821 | NP_898840.2 |
| AK034013.1 | 10 | 1785 | ||
| AK122448.2 | 6 | 876 | BAC65730.2 | |
| AK138598.1 | 10 | 1781 | ||
| AK163424.1 | 10 | 1803 | BAE37343.1 | |
| AK169957.1 | 10 | 1841 | BAE41480.1 | |
| BC139371.1 | 10 | 1804 | ||
| BC139397.1 | 10 | 1804 |
More Information
Set Membership: |
|

Genomic Map