Assay Details
Target Gene Details
Entrez Gene ID: | 381270 |
Gene Name: | membrane-associated ring finger (C3HC4) 4 |
Gene Aliases: |
BC056494, mKIAA1399 |
Location: |
Chr.1:72427112-72536564 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| March4 | NM_001045533.1 | 4 | 3238 | NP_001038998.1 |
| AK043443.1 | 1 | 508 | ||
| AK122502.1 | 4 | 3601 | BAC65784.1 | |
| BC056494.1 | 2 | 1141 | AAH56494.1 | |
| BC098191.1 | 4 | 3238 | AAH98191.1 |
More Information
Set Membership: |
|

Genomic Map