Assay Details
Target Gene Details
Entrez Gene ID: | 212679 |
Gene Name: | methionine-tRNA synthetase 2 (mitochondrial) |
Gene Aliases: |
C730026E21Rik, MetRS |
Location: |
Chr.1:55237177-55240058 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mars2 | NM_175439.3 | 1 | 953 | NP_780648.1 |
| AK042888.1 | 1 | 894 | BAC31393.1 | |
| AK050196.1 | 1 | 910 | BAC34119.1 | |
| AK144628.1 | 1 | 894 | ||
| AK172240.1 | 1 | 315 | BAE42903.1 | |
| BC099669.1 | 1 | 927 | AAH99669.1 | |
| BC132343.1 | 1 | 1006 | ||
| BC137739.1 | 1 | 1006 |
More Information
Set Membership: |
|

Genomic Map