Assay Details
Target Gene Details
Entrez Gene ID: | 17215 |
Gene Name: | minichromosome maintenance deficient 3 (S. cerevisiae) |
Gene Aliases: |
AL033361, C80350, Mcmd, P1, p1.m |
Location: |
Chr.1:20803014-20820213 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm3 | NM_008563.2 | 17 | 2568 | NP_032589.1 |
| AK088142.1 | 16 | 3074 | ||
| AK133850.1 | 17 | 2633 | BAE21885.1 | |
| AK145563.1 | 17 | 2667 | BAE26512.1 | |
| AK146497.1 | 17 | 2581 | BAE27213.1 | |
| AK147065.1 | 17 | 2583 | BAE27649.1 | |
| AK148006.1 | 17 | 2613 | BAE28283.1 | |
| AK195690.1 | 1 | 181 | ||
| AK219838.1 | 1 | 181 | ||
| BC031700.1 | 17 | 2568 | AAH31700.1 | |
| X62154.1 | 16 | 2512 | CAA44079.1 |
More Information
Set Membership: |
|

Genomic Map