Assay Details
Target Gene Details
Entrez Gene ID: | 214855 |
Gene Name: | AT rich interactive domain 5A (MRF1-like) |
Gene Aliases: |
D430024K22Rik, Mrf1 |
Location: |
Chr.1:36307706-36324036 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arid5a | NM_001172205.1 | 7 | 1320 | NP_001165676.1 |
| NM_001172206.1 | 7 | 1317 | NP_001165677.1 | |
| NM_001290726.1 | 8 | 1415 | NP_001277655.1 | |
| NM_001290727.1 | 6 | 1196 | NP_001277656.1 | |
| NM_145996.4 | 5 | 1146 | NP_666108.2 | |
| NR_033310.1 | 6 | 1199 | ||
| AK029237.1 | 5 | 1125 | ||
| AK041344.1 | 1 | 591 | BAC30913.1 | |
| AK085015.1 | 5 | 1126 | BAC39338.1 | |
| AK149839.1 | 7 | 1300 | BAE29115.1 | |
| AK150170.1 | 8 | 1395 | BAE29358.1 | |
| AK155555.1 | 6 | 1176 | ||
| AK156376.1 | 7 | 1300 | BAE33692.1 | |
| AK156622.1 | 6 | 1582 | BAE33779.1 | |
| AK156934.1 | 4 | 2882 | BAE33904.1 | |
| AK157607.1 | 7 | 1300 | BAE34133.1 | |
| BC027152.1 | 6 | 1163 | AAH27152.1 |
More Information
Set Membership: |
|

Genomic Map