Assay Details
Target Gene Details
Entrez Gene ID: | 68724 |
Gene Name: | ADP-ribosylation factor-like 8A |
Gene Aliases: |
1110033P22Rik, Arl10b, gie2 |
Location: |
Chr.1:135146834-135156268 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl8a | NM_026823.2 | 1 | 172 | NP_081099.1 |
| AB185207.1 | BAD30091.1 | |||
| BC018479.1 | 1 | 172 | AAH18479.1 |

Genomic Map