Assay Details
Target Gene Details
Entrez Gene ID: | 435653 |
Gene Name: | Fc receptor-like B |
Gene Aliases: |
FREB-2, FREB2, FcRL2, Fcrlm2, Fcry, mFCRL2 |
Location: |
Chr.1:170906852-170913860 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcrlb | NM_001029984.2 | 3 | 142 | NP_001025155.2 |
| AY513660.1 | 3 | 142 | AAS82875.1 | |
| DQ119292.1 | 3 | 142 | AAZ23789.1 |
More Information
Set Membership: |
|

Genomic Map