Assay Details
Target Gene Details
Entrez Gene ID: | 20343 |
Gene Name: | selectin, lymphocyte |
Gene Aliases: |
AI528707, CD62L, L-selectin, LECAM-1, Lnhr, Ly-22, Ly-m22, Lyam-1, Lyam1 |
Location: |
Chr.1:164061813-164080785 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sell | NM_001164059.1 | 8 | 1593 | NP_001157531.1 |
| NM_011346.2 | 9 | 1701 | NP_035476.1 | |
| AK137509.1 | 9 | 1613 | BAE23389.1 | |
| AK157273.1 | 9 | 1690 | ||
| AK163066.1 | 7 | 1158 | BAE37180.1 | |
| AK170753.1 | 8 | 1594 | BAE42004.1 | |
| BC052681.1 | 9 | 1685 | AAH52681.1 | |
| M25324.1 | 9 | 1693 | AAA39431.1 |

Genomic Map