Assay Details
Target Gene Details
Entrez Gene ID: | 100503816 |
Gene Name: | predicted gene 16548 |
Gene Aliases: |
- |
Location: |
Chr.1:164148576-164152217 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16548 | NR_037987.1 | 2 | 624 | |
| AK149537.1 | 2 | 397 |
Target Gene Details
Entrez Gene ID: | 20344 |
Gene Name: | selectin, platelet |
Gene Aliases: |
CD62P, GMP-140, Grmp, LECAM3, PADGEM |
Location: |
Chr.1:164115264-164150026 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selp | NM_011347.2 | 16 | 2792 | NP_035477.1 |
| AK145335.1 | 4 | 763 | ||
| M87861.1 | 15 | 2678 | AAA40008.1 |

Genomic Map