Assay Details
Target Gene Details
Entrez Gene ID: | 14261 |
Gene Name: | flavin containing monooxygenase 1 |
Gene Aliases: |
- |
Location: |
Chr.1:162829561-162866548 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fmo1 | NM_010231.2 | 9 | 1826 | NP_034361.1 |
| BC011229.1 | 9 | 1826 | AAH11229.1 | |
| D16215.1 | 9 | 1716 | BAA03745.1 | |
| U87456.1 | 9 | 1811 | AAB47569.1 |
More Information
Set Membership: |
|

Genomic Map