Assay Details
Target Gene Details
Entrez Gene ID: | 74106 |
Gene Name: | DDB1 and CUL4 associated factor 6 |
Gene Aliases: |
1200006M05Rik, Iqwd1, PC326 |
Location: |
Chr.1:165329500-165460553 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf6 | NM_028759.1 | 10 | 1567 | NP_083035.1 |
| AK004618.1 | 10 | 1430 | BAB23414.1 | |
| AK077238.1 | 1 | 85 | BAC36701.1 |
More Information
Set Membership: |
|

Genomic Map