Assay Details
Target Gene Details
Entrez Gene ID: | 226844 |
Gene Name: | major facilitator superfamily domain containing 7B |
Gene Aliases: |
9630055N22Rik, FLVCR, Flvcr1 |
Location: |
Chr.1:191005832-191026215 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd7b | NM_001081259.1 | 10 | 3306 | NP_001074728.1 |
| AK036315.1 | 9 | 2903 | ||
| AK044206.1 | 2 | 1686 | ||
| AK165872.1 | 10 | 3306 | ||
| BC024751.1 | 5 | 2284 |
More Information
Set Membership: |
|

Genomic Map