Assay Details
Target Gene Details
Entrez Gene ID: | 66615 |
Gene Name: | autophagy related 4B, cysteine peptidase |
Gene Aliases: |
2510009N07Rik, AW048066, Apg4b, Atg4bl, Autl1 |
Location: |
Chr.1:93755033-93789529 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg4b | NM_174874.3 | 13 | 2350 | NP_777363.1 |
| AK028712.1 | 13 | 2361 | BAC26079.1 | |
| AK031570.1 | 13 | 2351 | BAC27455.1 | |
| AK144239.1 | 13 | 2356 | BAE25791.1 |
More Information
Set Membership: |
|

Genomic Map