Assay Details
Target Gene Details
Entrez Gene ID: | 545389 |
Gene Name: | centrosomal protein 170 |
Gene Aliases: |
4933426L22Rik, A330004A13Rik, AI195353 |
Location: |
Chr.1:176733653-176814173 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep170 | NM_001099637.2 | 20 | 4992 | NP_001093107.2 |
| AK016937.1 | BAB30507.2 | |||
| AK038878.1 | 7 | 1561 | ||
| AK078541.1 | 3 | 474 | BAC37328.1 | |
| AK172953.1 | 8 | 2501 | BAD32231.1 |
More Information
Set Membership: |
|

Genomic Map