Assay Details
Target Gene Details
Entrez Gene ID: | 213452 |
Gene Name: | dual serine/threonine and tyrosine protein kinase |
Gene Aliases: |
A930019K20Rik, C430014H23Rik, C820013G01, Ripk5 |
Location: |
Chr.1:132417181-132466959 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dstyk | NM_172516.4 | 13 | 5173 | NP_766104.2 |
| AK044117.1 | 1 | 257 | ||
| AK050542.1 | 9 | 3419 | BAC34316.1 | |
| AK082434.1 | 1 | 252 | ||
| AK082904.1 | 1 | 350 | ||
| AK145449.1 | BAE26444.1 | |||
| AK172954.1 | 13 | 5019 | BAD32232.1 | |
| AY429676.1 | 13 | 5059 | AAS55392.1 | |
| BC089380.1 | 10 | 3590 | AAH89380.1 |
More Information
Set Membership: |
|

Genomic Map