Assay Details
Target Gene Details
Entrez Gene ID: | 67769 |
Gene Name: | G patch domain containing 2 |
Gene Aliases: |
5830433G22Rik, 5830436K05Rik, AI427714, AI447508, AW107440, AW491060, Gpatc2 |
Location: |
Chr.1:187215393-187369480 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpatch2 | NM_026367.4 | 2 | 764 | NP_080643.4 |
| AK017975.1 | 2 | 733 | BAB31021.1 | |
| AK029990.1 | 2 | 749 | ||
| AK030026.1 | 2 | 763 | BAC26744.1 | |
| AK032734.1 | 2 | 739 | ||
| AK053781.1 | 2 | 756 | BAC35520.1 | |
| AK167826.1 | 2 | 768 | BAE39848.1 | |
| AK169660.1 | 2 | 765 | ||
| BC054810.1 | 2 | 736 | AAH54810.1 |
More Information
Set Membership: |
|

Genomic Map