Assay Details
Target Gene Details
Entrez Gene ID: | 240888 |
Gene Name: | G protein-coupled receptor 161 |
Gene Aliases: |
Gm208, Gm208Gpr, vl |
Location: |
Chr.1:165281174-165326746 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr161 | NM_001081126.1 | 4 | 700 | NP_001074595.1 |
| AK145635.1 | 3 | 724 | ||
| AY255596.1 | 2 | 191 | AAO85108.1 | |
| BC137659.1 | 3 | 638 | ||
| EF197953.1 | 3 | 725 |
More Information
Set Membership: |
|

Genomic Map