Assay Details
Target Gene Details
Entrez Gene ID: | 240888 |
Gene Name: | G protein-coupled receptor 161 |
Gene Aliases: |
Gm208, Gm208Gpr, vl |
Location: |
Chr.1:165281174-165326746 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr161 | NM_001081126.1 | 7 | 1863 | NP_001074595.1 |
| AK145635.1 | 6 | 1888 | ||
| BC137659.1 | ||||
| EF197953.1 | 6 | 1888 |
More Information
Set Membership: |
|

Genomic Map