Assay Details
Target Gene Details
Entrez Gene ID: | 171469 |
Gene Name: | G protein-coupled receptor 37-like 1 |
Gene Aliases: |
AW047233, CAG-18, D0Kist8 |
Location: |
Chr.1:135160250-135167681 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr37l1 | NM_134438.3 | 2 | 1771 | NP_602320.2 |
| AB016602.1 | 2 | 1514 | BAA32062.1 | |
| AK019303.1 | ||||
| AK158275.1 | 2 | 1773 | ||
| BC098482.1 | 2 | 1462 | ||
| BC127619.1 | 2 | 1395 | ||
| BC127620.1 | 2 | 1395 | ||
| CD774808.1 | 1 | 497 | ||
| U80895.1 | 1 | 825 |
More Information
Set Membership: |
|

Genomic Map