Assay Details
Target Gene Details
Entrez Gene ID: | 57814 |
Gene Name: | potassium voltage-gated channel, Isk-related subfamily, gene 4 |
Gene Aliases: |
AW487937, MiRP3 |
Location: |
Chr.1:78816949-78820025 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcne4 | NM_021342.1 | 2 | 852 | NP_067317.1 |
| AF076533.1 | 2 | 852 | AAD28091.1 | |
| AK133469.1 | 2 | 891 | ||
| BC032920.1 | 2 | 880 | AAH32920.1 | |
| BC034849.1 | AAH34849.1 |
More Information
Set Membership: |
|

Genomic Map