Assay Details
Target Gene Details
Entrez Gene ID: | 69163 |
Gene Name: | mitochondrial ribosomal protein L44 |
Gene Aliases: |
1810030E18Rik, 5730593H20Rik, AI463302 |
Location: |
Chr.1:79776018-79781445 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl44 | NM_001081210.1 | 4 | 1106 | NP_001074679.1 |
| AK007652.1 | 3 | 859 | ||
| AK146024.1 | 4 | 918 | BAE26840.1 | |
| AK150537.1 | 4 | 868 | BAE29642.1 | |
| AK151854.1 | 4 | 868 | BAE30743.1 | |
| AK156814.1 | 4 | 1021 | BAE33863.1 | |
| BC019727.1 | 2 | 339 | AAH19727.1 |
More Information
Set Membership: |
|

Genomic Map