Assay Details
Target Gene Details
Entrez Gene ID: | 104009 |
Gene Name: | quiescin Q6 sulfhydryl oxidase 1 |
Gene Aliases: |
1300003H02Rik, QSOX, Qscn6, SOx |
Location: |
Chr.1:155778155-155812899 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qsox1 | NM_001024945.1 | 12 | 3252 | NP_001020116.1 |
| NM_023268.2 | 13 | 2386 | NP_075757.1 | |
| AK004880.1 | 13 | 2332 | BAB23638.1 | |
| AK009444.1 | 1 | 523 | ||
| AK012943.1 | 3 | 931 | ||
| AK083938.1 | 12 | 3203 | BAC39073.1 | |
| AK166839.1 | 12 | 3198 | BAE39061.1 | |
| AK169877.1 | 13 | 2618 | BAE41429.1 | |
| AK170449.1 | 13 | 2299 | BAE41807.1 | |
| AK211239.1 | ||||
| BC034131.1 | 10 | 1878 | AAH34131.1 | |
| BC076590.1 | 12 | 3206 | AAH76590.1 | |
| CK625426.1 | 1 | 148 |
More Information
Set Membership: |
|

Genomic Map