Assay Details
Target Gene Details
Entrez Gene ID: | 69527 |
Gene Name: | mitochondrial ribosomal protein S9 |
Gene Aliases: |
2310002A08Rik, AW322516, Mrp-S9, S9mt, mg637 |
Location: |
Chr.1:42851233-42905683 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps9 | NM_023514.4 | 11 | 1185 | NP_076003.3 |
| AJ250193.1 | CAC08499.1 | |||
| AK009082.1 | 11 | 1135 | BAB26060.2 | |
| AK150985.1 | 11 | 1142 | ||
| BC022587.1 | 6 | 623 | AAH22587.1 | |
| BC051936.1 | 10 | 878 | AAH51936.1 | |
| BC069980.1 | 9 | 779 | AAH69980.1 | |
| BC116437.2 | 11 | 1135 | ||
| DQ700381.1 |
More Information
Set Membership: |
|

Genomic Map