Assay Details
Target Gene Details
Entrez Gene ID: | 226499 |
Gene Name: | cDNA sequence BC003331 |
Gene Aliases: |
1810053E15Rik, Odr4, odr-4 |
Location: |
Chr.1:150361311-150393055 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC003331 | NM_001077237.1 | 1 | 179 | NP_001070705.1 |
| NM_145511.2 | 1 | 179 | NP_663486.2 | |
| BY247764.1 | 1 | 179 |
Target Gene Details
Entrez Gene ID: | 108989 |
Gene Name: | translocated promoter region, nuclear basket protein |
Gene Aliases: |
2610029M07Rik, C77892 |
Location: |
Chr.1:150392838-150449935 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpr | NM_133780.3 | NP_598541.3 | ||
| AK076012.1 | BAC36119.1 |
More Information
Set Membership: |
|

Genomic Map