Assay Details
Target Gene Details
Entrez Gene ID: | 56406 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181 |
Location: |
Chr.2:155390656-155473894 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa6 | NM_001242558.1 | NP_001229487.1 | ||
| NM_019825.3 | 10 | 4065 | NP_062799.2 | |
| AF216186.1 | 9 | 3950 | AAF35860.1 | |
| AK129079.1 | 3 | 1224 | BAC97889.1 | |
| AK142566.1 | 4 | 2250 | BAE25107.1 | |
| AK171864.1 | 10 | 4223 | BAE42705.1 | |
| BC083187.1 | 10 | 4065 | AAH83187.1 | |
| JF707634.1 | ||||
| JF707635.1 | 9 | 3924 |
More Information
Set Membership: |
|

Genomic Map