Assay Details
Target Gene Details
Entrez Gene ID: | 56406 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181 |
Location: |
Chr.2:155390656-155473894 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa6 | NM_001242558.1 | 13 | 3715 | NP_001229487.1 |
| NM_019825.3 | 14 | 6691 | NP_062799.2 | |
| AF216186.1 | 13 | 6576 | AAF35860.1 | |
| AK129079.1 | 7 | 3850 | BAC97889.1 | |
| AK136585.1 | 4 | 1463 | ||
| AK142566.1 | 8 | 4876 | BAE25107.1 | |
| AK161598.1 | 2 | 1491 | ||
| BC083187.1 | 14 | 6691 | AAH83187.1 | |
| CJ056778.1 | 1 | 429 |
More Information
Set Membership: |
|

Genomic Map