Assay Details
Target Gene Details
Entrez Gene ID: | 228005 |
Gene Name: | peptidyl-prolyl isomerase G (cyclophilin G) |
Gene Aliases: |
AU019516, AU022200, B230312B02Rik, CYP, SRCyp |
Location: |
Chr.2:69722617-69754059 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppig | NM_001081086.1 | 1 | 226 | NP_001074555.1 |
| AK088007.1 | ||||
| AK134121.1 | 1 | 227 | BAE22023.1 | |
| AK135178.1 | 1 | 268 | ||
| AK163324.1 | ||||
| BC016126.1 | ||||
| BC027420.1 | 1 | 221 | ||
| BC048905.1 | AAH48905.1 | |||
| BC094926.1 | AAH94926.1 | |||
| BC150694.1 | 1 | 248 | ||
| U91923.1 | AAC05726.1 |
More Information
Set Membership: |
|

Genomic Map